Human Recombinant Annexin V (from E. coli), APC (Allophycocyanin)
Supplier: Enzo Life Sciences
209-252-T020
209-252-T100
89145-078EA
252.8
CAD
89145-078
89145-080
Human Recombinant Annexin V (from E. coli), APC (Allophycocyanin)
Proteins and Peptides
Produced in E. coli.
APC-labelled recombinant human annexin V is suitable for dual staining experiments to detect phosphatidylserine whilst cell-surface protein can be detected with a suitable MAb (FITC-labelled or labelled secondary antibody). The fluorochrome APC (allophycocyanin) shows far red fluorescence (Ex(max): 650nm; Em(max): 660nm).
Ordering information: Quantity: 20 tests/100µl; 100 tests/500µl.