Human Recombinant Caspase-9 (from E. coli)

Supplier: Enzo Life Sciences

201-047-U025 201-047-U100
89143-996EA 630.42 CAD
89143-996 89143-998
Human Recombinant Caspase-9 (from E. coli)
Enzymes

Produced in E. coli. Contains an N-terminal His-tag.

Order Now

SPECIFICATIONS

Enzyme name Caspase-9
Enzyme synonyms ICE-LAP-6
Mch6
Environmentally Preferable
Enzyme type Recombinant
Source E. coli
Species Human
Form Lyophilized
Purity ≥90% (SDS-PAGE)
Storage conditions –80 °C
Reconstitution instructions Reconstitute to 1 U/µl with 25 µl or 100 µl PBS containing 15% glycerol.
Note: It is normal to see some insoluble protein following reconstitution. This is mainly due to denaturation of small amounts of protein during preparation. Precipitate should not affe
Enzyme activity ~400 U/mg protein. One unit is defined as the amount of enzyme that cleaves 1 nmol of the caspase substrate LEHD-pNA per hour at 37 °C in a reaction solution containing 50 mM HEPES, pH 7.2, 50 mM NaCl, 0.1% CHAPS, 10 mM EDTA, 5% glycerol and 10 mM DTT.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR