Recombinant N-Acetyl-eglin C
Supplier: Enzo Life Sciences
201-006-M001
201-006-MC05
89143-966EA
1638.46
CAD
89143-966
89143-968
89143-970
Recombinant N-Acetyl-eglin C
Proteins and Peptides
Protease inhibitor
Eglin C was originally isolated from the leech Hirudo medicinalis . Recombinant N-acetyl-eglin C is a 70 amino acid peptide with identical biological activity to native eglin C. An effective inhibitor of chymotrypsin and subtilisin as well as of leukocyte elastase and cathepsin G.