Substance P
Supplier: Enzo Life Sciences
167-003-M001
167-003-M005
89143-830EA
88.48
CAD
89143-830
89143-832
Substance P
Proteins and Peptides
Neuropeptide
Neuropeptide inducing Ca2+ mobilization. Activates G1-Proteins directly. Induces superoxide production in neutrophils.