Melittin
Supplier: Enzo Life Sciences
162-006-M001
162-006-M005
89143-538EA
143.78
CAD
89143-538
89143-540
Melittin
Proteins and Peptides
PLA2 activator
Causes disruption of normal cellular activity and cell lysis. Binds calmodulin in a calcium-dependent manner. Activates phospholipase A2 (PLA2) and inhibits protein kinase C (PKC) by binding to the catalytic domain in a Mg-ATP sensitive manner. Antirheumatic.