Human Humanin
Supplier: Enzo Life Sciences
158-011-M001 158-011-MC05
89143-468EA 584.6 CAD
89143-468 89143-470
Human Humanin
Proteins and Peptides
Bax inhibitor
Humanin was originally identified as an anti-apoptotic peptide of 24 aa encoded in a cDNA that rescued neuronal cells from apoptosis induced by presenilin mutants associated with familial Alzheimer's disease and by amyloid-beta protein. Mutagenesis studies showed that the cysteine at position 8 of the humanin peptide is critical for its anti-apoptotic function. It has now been shown that Bax (Bcl-2 associated X protein), an apoptosis-inducing protein, interacts with humanin. Humanin prevents the translocation of Bax from the cytosol to mitochondria. Humanin peptides also block Bax association with isolated mitochondria, and suppress cytochrome c release in vitro.