Caveolin-1 Scaffolding Domain Peptide
Supplier: Enzo Life Sciences
153-064-M001 153-064-M005
89143-236EA 372.88 CAD
89143-236 89143-238
Caveolin-1 Scaffolding Domain Peptide
Proteins and Peptides
Cell permeable peptide that blocks eNOS (NOS III) activity and cellular nitric oxide (NO) release in vitro and reduces inflammation and tumorigenesis in vivo. Caveolin-1 interacts with several lipid-modified signalling ligands, such as EGFR, eNOS, G-protein α-subunits, PKCα, H-Ras, and Src, via the C1-SD82-101 sequence.