N-Acetyl-Asp-Glu-OH
Supplier: Enzo Life Sciences
151-020-M010 151-020-M050
89143-004EA 115.34 CAD
89143-004 89143-006
N-Acetyl-Asp-Glu-OH
Proteins and Peptides
Glutamate receptor ligand
Endogenous neurotransmitter with high affinity for the brain glutamate receptor. N-acetylated α-linked acidic dipeptidase (NAALADase), a membrane-bound peptidase, hydrolyzes α-NAAG, a major brain peptide, to N-acetylaspartate and glutamate. Discriminates between mGluR2 and mGluR3. mGluR3 activator. NMDA receptor antagonist. Neuroprotective.