L-Thiocitrulline dihydrochloride ≥97%
Supplier: Enzo Life Sciences
106-013-M010 106-013-M050
89142-870EA 121.66 CAD
89142-870 89142-872
L-Thiocitrulline dihydrochloride ≥97%
L-Thiocitrulline dihydrochloride
NOS inhibitor
Inhibitor of neuronal (nNOS; NOS I; Ki=60nM) and inducible (iNOS; NOS II) nitric oxide synthase.
CAS Number: 212051-53-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c47704dc56ba9c94ce075e09668bf90bb879b62&vid=044aaea4a2963043961ef6b53e91d57197782260&prid=web&clid=SAPDAM