Nω-Methyl-L-arginine monoacetate ≥99% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: H-Arg(Me)-OH.AcOH, N5-(Methylamidino)-L-ornithine acetate salt, Nomega-Monomethyl-L-arginine acetate, L-NMMA, Ng-Methyl-L-Arginine Acetate Salt, Nomega-Methyl-L-arginine acetate, N-ω-Monomethyl-L-arginine monoacetate, NG-Monomethyl-L-arginine acetate salt

106-001-G001 106-001-M025 106-001-M100
89142-822EA 703.1 CAD
89142-822 89142-826 89142-828
Nω-Methyl-L-arginine monoacetate ≥99% (by TLC)
Nω-Methyl-L-arginine monoacetate

Competitively inhibits the generation of NO from arginine and is a useful tool for inhibition of NO mediated effects. In vitro, L-NMMA inhibits relaxation of rat aorta rings induced by acetylcholine (IC50=9.5µM). In vivo, it causes a dose dependent increase in mean arterial blood pressure in anesthetized rats (EC50=16.7mg/kg).


  • NOS inhibitor


White to off-white solid.


Soluble in water, 100% ethanol (1mg/ml) or DMSO (0.25mg/ml).

Formula: C₇H₁₆N₄O₂·CH₃COOH
Storage Temperature: Refrigerator
MDL Number: MFCD00069311
CAS Number: 53308-83-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee9de664dccbbe960cd1174818f114119469f4d1&vid=748145bfa865a0dc31c3695387cfb7939dfa8732&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR