Nω-Nitro-L-arginine methyl ester hydrochloride ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: N-ω-Nitro-L-Arginine Methyl Ester Hydrochloride, L-NAME Hydrochloride, L-NAME HCl, H-Arg(NO2)-OMe.HCl, (S)-Methyl 2-amino-5-(3-nitroguanidino)pentanoate hydrochloride

105-003-G005 105-003-G025
89142-800EA 146.94 CAD
89142-800 89142-802
Nω-Nitro-L-arginine methyl ester hydrochloride ≥98% (by TLC)
Nomega-Nitro-L-arginine methyl ester hydrochloride

Inhibitor of nitric oxide synthase (NOS). More soluble than NG-Nitro-L-arginine. Suppresses the release of nitric oxide (NO) from endothelial cells. Has a different effect on the superoxide generation by NOS than L-NMMA.


  • NOS inhibitor


White to off-white solid.


Soluble in water (50mg/ml) or methanol (25g/ml).

Formula: C₇H₁₅N₅O₄
MW: 269.69 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00039052
CAS Number: 51298-62-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3ba5f1c0156c9796837178b038b47ec0613858c&vid=ef085ead03bbf508bb6596b4691eb4e43143ee9d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR