Hamster Recombinant Grp78/BiP (from E. coli), Enzo Life Sciences

Supplier: Enzo Life Sciences

ADI-SPP-765-D ADI-SPP-765-F
89142-256EA 455.04 CAD
89142-256 89142-258
Hamster Recombinant Grp78/BiP (from E. coli), Enzo Life Sciences
Proteins and Peptides

Produced in E. coli.

Order Now

SPECIFICATIONS

Protein/peptide name Grp78 Grp78
Environmentally Preferable
Protein synonyms BiP BiP
Protein/peptide type Recombinant Recombinant
Species Hamster Hamster
Source E. coli E. coli
Purity >80% (SDS-PAGE; Western blot)
Form Liquid
Formulation In TRIS buffer, pH 7.5, containing sodium chloride, glycerol, and DTT.
Purification Purified by multi-step chromatography.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR