Human Recombinant HO-1 (from E. coli)

Supplier: Enzo Life Sciences

ADI-SPP-732-D ADI-SPP-732-F
89142-224EA 532.46 CAD
89142-224 89142-226
Human Recombinant HO-1 (from E. coli)
Proteins and Peptides

Produced in E. coli.

Order Now

SPECIFICATIONS

Protein/peptide name HO-1 HO-1
Environmentally Preferable
Protein/peptide type Recombinant Recombinant
Species Human Human
Source E. coli E. coli
Purity >90% (SDS-PAGE; Western blot)
Form Liquid
Formulation In 50mM TRIS-HCl, pH 7.5, containing 2% glycerol, 450mM sodium chloride, 1mM DTT, and 0.1mM PMSF.
Purification Purified by multi-step chromatography.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR