Anti-HDJ1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-400-D ADI-SPA-400-F
89142-048EA 298.62 CAD
89142-048 89142-050
Anti-HDJ1 Rabbit Polyclonal Antibody
Antibodies
Hsp40/Hdj1 is a cytosolic co-chaperone belonging to the class DnaJ, named after its homolog in E. coli. Hsp40 functions in protein folding by binding nascent peptides and unfolded substrates and facilitating substrate interaction with Hsp70. Binding of Hsp40 to Hsp70 increases Hsp70 ATP hydrolysis and substrate binding, which is reversed by the action of nucleotide exchange factors (GrpE in E. coli). Repeated cycles of Hsp40/Hsp70 peptide binding and ATP hydrolysis prevent premature folding and aggregation, promoting protein maturation throughout the cell.

  • For Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, beluga, dog, chicken, fish, guinea pig, hamster, mouse, monkey, mussel, pig, rabbit, sheep, scallop and Xenopus HSP40 (Hdj1). Detects a band of ~40kDa by Western blot.

Type: Primary
Antigen: HDJ1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR