Anti-NF-kappa B P65 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAS-TF110-D KAS-TF110-F
89135-314EA 346.02 CAD
89135-314 89135-316
Anti-NF-kappa B P65 Rabbit Polyclonal Antibody
Antibodies
Nuclear factor kappa-B (NFk) B) functions as a sequence specific transcriptional activator that binds to the intronic enhancer of kappa light chain gene in B lymphocytes. NF-kB is a heterodimer consisting of a 50 kDa DNA binding subunit (p50) and a 65 kDa transactivation subunit (p65/RelA) (ref. 3), both of which exhibit sequence homology to the protooncogene c-Rel. p50 has an isoform called p49/p52, and both proteins derive from the amino-terminal of precursor protein p105 and p100, respectively. The p50/p65 heterodimer remains in inactive form in the cytosol as a complex with its inhibitor, IkB. Upon stimulation of cells by a variety of stimuli, including lipopolysaccharide (LPS), pro-inflammatory cytokines like interleukin-1 and tumor necrosis factor, and viral infection, IkB becomes phosphorylated and degraded by the proteosome. The active NF-kB heterodimer translocates into the nucleus and induces gene expression.

  • For Western Blot

Recognizes human, mouse, rat, bovine, dog, guinea pig, monkey, pig and rabbit NF-kappaB p65. Detects a band of ~65kDa by Western blot.

Type: Primary
Antigen: NF-KAPPAB P65
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR