Anti-RAS Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-GP001-D KAP-GP001-F
89133-390EA 331.8 CAD
89133-390 89133-392
Anti-RAS Rabbit Polyclonal Antibody
Antibodies
Monomeric G-protein Ras functions as a molecular switch linking receptor and nonreceptor tyrosine kinase activation to a number of downstream signaling pathways. Oncogenic mutations, which occur in approximately 30% of all cases of human cancer, lead to the stimulation of proliferation and the inhibition of apoptosis.

  • For Western Blot

Recognizes human, mouse, rat and bovine Ras. Detects a band of 21-23kDa by Western blot.

Type: Primary
Antigen: RAS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR