17β-Estradiol High Sensitivity ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-174 ADI-901-174
89141-256EA 646.22 CAD
89141-256 89141-346
17β-Estradiol High Sensitivity ELISA Kit, Enzo Life Sciences
Assays ELISAs
The 17beta-Estradiol (Serum/Plasma) EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours.
  • Highly sensitive measurement
  • Fast and reliable results
  • ully quantitative results that surpass semi-quantitative Western blot analysis
  • Convenient pre coated plate and color coded reagents reduces errors
For the quantitative determination of 17beta-Estradiol in plasma and serum from any species.
Delivery information: Kit includes DxS microtiter plate, conjugate, antibody, assay buffer concentrate, wash buffer concentrate, standard, pNpp substrate, stop solution.