Aldosterone ELISA kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-901-173 ADI-900-173
89141-254EA 636.99 CAD
89141-254 89141-344
Aldosterone ELISA kit, Enzo Life Sciences
Assays ELISAs
The Aldosterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour.
  • Highly sensitive
  • Negligible reactivity with other steroids
  • Fully quantitative results that surpass semi-quantitative Western blots
For the quantitative determination of Aldosterone in plasma, serum, and urine from any species.
Delivery information: Kit includes DxS microtiter plate, conjugate, antibody, assay buffer concentrate, wash buffer concentrate, standard, pNpp substrate, stop solution.