Corticosterone ELISA kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-901-097 ADI-900-097
89141-146EA 645.7 CAD
89141-146 101106-156
Corticosterone ELISA kit, Enzo Life Sciences
Assays ELISAs
The Corticosterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours
  • Sensitive measurement of Corticosterone
  • High throughput results
  • Routinely used to monitor stress levels in zoo animals
This is widely used sensitive corticosterone ELISA kit with a large variety of sample types and highly reproducible results for animal stress research. For the quantitative determination of Corticosterone in culture supernatants, feces, plasma, serum, and saliva from any species. Cited sample types include cell lysate and whole blood.
Absorbance is read at 405 nm. The Corticosterone EIA kit has low cross-reactivity with related steroids. The broad dynamic range makes this kit ideal for a wide variety of sample matrices from any species
Ordering information: Alternative size available as ADI-900-097 (1x96 wells)
Delivery information: Kit includes DxS microtiter plate, conjugate, antibody, assay buffer 15 concentrate, wash buffer concentrate, standard, pNpp substrate, stop solution, steroid displacement reagent.