Sulfo-GMBS

Supplier: BACHEM AMERICAS INC

4077463.0050 4077463.0100
Q-2770.0050BAEA 513.39 CAD
Q-2770.0050BA Q-2770.0100BA
Sulfo-GMBS
Sulfo-GMBS

Sequence: Sulfo-N-succinimidyl 4-maleimidobutyrate sodium salt
Synonym(s): Sulfo-GMBS sodium salt


Sulfo-N-succinimidyl 4-maleimidobutyrate (Sulfo-GMBS) is a water-soluble, heterobifunctional crosslinking reagent for conjugation of cysteine-containing peptides to carrier proteins or surfaces.

Formula: C₁₂H₁₁N₂NaO₉S
Storage Temperature: Freezer
MDL Number: MFCD17215919
CAS Number: 185332-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR