2-((5-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-10,11-dihydro-5H-dibenzo[a,d][7]annulen-2-yl)oxy)acetic acid
Supplier: BACHEM AMERICAS INC
Synonyms:
Ramage Linker, Fmoc-Suberol
4025359.0005
4025359.0025
Q-2745.0005BAEA
377.3
CAD
Q-2745.0005BA
Q-2745.0025BA
2-((5-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-10,11-dihydro-5H-dibenzo[a,d][7]annulen-2-yl)oxy)acetic acid
2-((5-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-10,11-dihydro-5H-dibenzo[a,d][7]annulen-2-yl)oxy)acetic acid
Sequence: 5-Fmoc-amino-2-carboxymethoxy-10,11-dihydro-5H-dibenzo[a,d]cycloheptene
Synonym(s): Ramage Linker
A linker suitable for Fmoc-SPPS of peptide amides. Cleavage is achieved e.g. with 95% TFA, scavengers may be required. Bachem also offers this linker bound to polystyrene/1% DVB resin, D-2200.
Formula:
C₃₂H₂₇NO₅ MW: 505.6 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07783960 CAS Number: 212783-75-0 |
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...