6-Azidohexanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: 6-Azido-hexanoic acid

4066756.0001 4066756.0005
Q-2740.0001BAEA 774.5 CAD
Q-2740.0001BA Q-2740.0005BA
6-Azidohexanoic acid
6-Azidohexanoic acid

Sequence: 6-Azido-hexanoic acid
Synonym(s): ε-Azidocaproic acid


Buiding blocks for "click chemistry". See also the product family "Propargylglycine" and the subfamily "p-Azidophenylalanine". Bachem additionally offers azido derivatives for SPPS: Fmoc-Dab(N₂)-OH (B-4145), Fmoc-Orn(N₂)-OH (B-4140), Fmoc-Lys(N₂)-OH (B-4135), Boc-Orn(N₂)-OH · DCHA (A-4770), and Boc-Lys(N₂)-OH · DCHA (A-4765).


Building block for Staudinger ligation as well as for "click chemistry". This highly selective reaction can be applied for the conjugation of peptides with various molecules and for the cyclization of peptides. The azido moiety may also be considered as a protected amino group which is deblocked under reductive conditions.

Formula: C₆H₁₁N₃O₂
MW: 157.173 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12406156
CAS Number: 79598-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR