COMU
Supplier: BACHEM AMERICAS INC
Synonyms:
4-(6-Cyano-2-methyl-7-oxo-4,8-dioxa-2,5-diazadec-5-en-3-ylidene)morpholin-4-ium hexafluorophosphate(V), (1-Cyano-2-ethoxy-2-oxoethylidenaminooxy)dimethylamino-morpholino-carbenium hexafluorophosphate
Sequence: 1-[1-(Cyano-2-ethoxy-2-oxoethylideneaminooxy)-dimethylamino-morpholino]-uronium hexafluorophosphate
A safe and highly efficient coupling reagent requiring only one equivalent of base during bond formation as it liberates morpholine. This helps to minimize concomitant racemization. Non-allergenic. Couplings with COMU can be monitored visually or colorimetrically, the reagent shows excellent solubility in DMF and NMP. COMU is especially suited for microwave-accelerated SPPS.
Formula:
C₁₂H₁₉F₆N₄O₄P MW: 428.272 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD11975052 CAS Number: 1075198-30-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...