5-Carboxyfluorescein
Supplier: BACHEM AMERICAS INC
Synonyms:
Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-5-carboxylic acid, 3',6'-dihydroxy-3-oxo-, 5-FAM, 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylic acid, 5-Carboxyfluorescein hydrate
Sequence: 5-Carboxy-fluorescein
Synonym(s): 5-FAM#4-(6-Hydroxy-3-oxo-3H-xanthen-9-yl)-isophthalic acid
Bachem also offers the coumarin derivative Fmoc-3-(7-methoxy-coumarin-4-yl)-Ala-OH (B-3740), the (7-Hydroxycoumarin-4-yl)ethylglycine derivatives B-4205 and F-4270, and a range of Nε-labeled lysine derivatives (please see the subfamily "Nε-Labeled Lysine").
Widely used green-fluorescent labeling dye.
Formula:
C₂₁H₁₂O₇.xH₂O MW: 376.32 g/mol (anhy) Storage Temperature: Freezer |
MDL Number:
MFCD00036874 CAS Number: 76823-03-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...