1-Methylimidazole-2-carboxylic acid

Supplier: BACHEM AMERICAS INC

4035264.0001 4035264.0005
Q-2605.0001BAEA 258.14 CAD
Q-2605.0001BA Q-2605.0005BA
1-Methylimidazole-2-carboxylic acid
1-Methylimidazole-2-carboxylic acid

Sequence: 1-Methyl-1H-imidazole-2-carboxylic acid

Formula: C₅H₆N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD01863421
CAS Number: 20485-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR