3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one

Supplier: BACHEM AMERICAS INC

4029249.0025 4029249.0100
Q-2565.0025BAEA 619.6 CAD
Q-2565.0025BA Q-2565.0100BA
3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one
3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one

Sequence: 3-(Diethoxy-phosphoryloxy)-1,2,3-benzo[d]triazin-4(3H)-one


DEPBT, a crystalline phosphonium-type coupling reagent, mediates amide bond formation with a remarkable resistance to racemization. Thus, DEPBT is an excellent activation reagent for coupling the racemization-prone Fmoc-His(Trt)-OH. The use of DEPBT permitted the total synthesis of the antiviral peptide antibiotic feglymycin, which contains a high proportion of ring-substituted phenylglycines.

Formula: C₁₁H₁₄N₃O₅P
Storage Temperature: Freezer
MDL Number: MFCD01236967
CAS Number: 165534-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR