SPDP (N-Succinimidyl-3-(2-pyridyldithio)propionate)

Supplier: BACHEM AMERICAS INC

Synonyms: 2,5-Dioxopyrrolidin-1-yl 3-(pyridin-2-yldisulphanyl)propanoate, 3-(2-Pyridyldithio)propionic Acid N-Succinimidyl Ester, SPDP, 2,5-Dioxopyrrolidin-1-yl 3-(pyridin-2-yldisulfanyl)propanoate, 3-(2-Pyridyldithio)-Propionic Acid N-Hydroxysuccinimide Ester

4029094.0250 4029094.1000
Q-2535.0250BAEA 1032.37 CAD
Q-2535.0250BA Q-2535.1000BA
SPDP (N-Succinimidyl-3-(2-pyridyldithio)propionate)
SPDP (N-Succinimidyl-3-(2-pyridyldithio)propionate)

Sequence: 3-(2-Pyridyldithio)-propionic acid-OSu
Synonym(s): 3-(Pyridin-2-yldisulfanyl)-propionic acid-OSu#SPDP


Important crosslinking reagent for the preparation of protein-protein and peptide-protein conjugates linked by disulfide bonds.

Formula: C₁₂H₁₂N₂O₄S₂
Storage Temperature: Freezer
MDL Number: MFCD00009636
CAS Number: 68181-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR