Benzyl-N-(4-aminobutyl)carbamate

Supplier: BACHEM AMERICAS INC

Synonyms: N-1-Z-1,4-diaminobutane·HCl

4028903.0001 4028903.0005
Q-2515.0001BAEA 309.81 CAD
Q-2515.0001BA Q-2515.0005BA
Benzyl-N-(4-aminobutyl)carbamate
Benzyl-N-(4-aminobutyl)carbamate

Sequence: N-1-Z-1,4-diaminobutane · HCl


Mono-protected alkylene diamines are important homobifunctional compounds often used as spacer groups (especially hexamethylenediamine), e.g. for the immobilization of peptides. Additionally they are very useful building blocks for preparing polyamines and polyamides. 1,5-Diaminopentane (cadaverine) is the decarboxylation product of lysine, ornithine yields 1,4-diaminobutane (putrescine).

Formula: C₁₂H₁₈N₂O₂
MW: 222.288 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00270149
CAS Number: 62146-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR