9H-Fluoren-9-ylmethyl-N-(4-aminobutyl)carbamate

Supplier: BACHEM AMERICAS INC

4027909.0001 4027909.0005
Q-2420.0001BAEA 309.81 CAD
Q-2420.0001BA Q-2420.0005BA
9H-Fluoren-9-ylmethyl-N-(4-aminobutyl)carbamate
9H-Fluoren-9-ylmethyl-N-(4-aminobutyl)carbamate

Sequence: N-1-Fmoc-1,4-diaminobutane · HCl


Mono-protected alkylene diamines are important homobifunctional compounds often used as spacer groups (especially hexamethylenediamine), e.g. for the immobilization of peptides. Additionally they are very useful building blocks for preparing polyamines and polyamides. 1,5-Diaminopentane (cadaverine) is the decarboxylation product of lysine, ornithine yields 1,4-diaminobutane (putrescine).

Formula: C₁₉H₂₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD01631652
CAS Number: 117048-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR