N-(tert-Butoxycarbonyl)-1,5-diaminopentane

Supplier: BACHEM AMERICAS INC

Synonyms: N-Boc-1,5-diaminopentane, tert-Butyl N-(5-Aminopentyl)carbamate, tert-Butyl (5-aminopentyl)carbamate, N-(5-Aminoamyl)carbamic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)-1,5-pentanediamine, N-(5-Aminopentyl)carbamic Acid tert-Butyl Ester, NH2-C5-NH-Boc, N-Boc-1,5-pentanediamine

4027865.0001 4027865.0005
Q-2380.0001BAEA 317.74 CAD
Q-2380.0001BA Q-2380.0005BA
N-(tert-Butoxycarbonyl)-1,5-diaminopentane
N-(tert-Butoxycarbonyl)-1,5-diaminopentane

Sequence: N-1-Boc-1,5-diaminopentane · HCl


Mono-protected alkylene diamines are important homobifunctional compounds often used as spacer groups (especially hexamethylenediamine), e.g. for the immobilization of peptides. Additionally they are very useful building blocks for preparing polyamines and polyamides. 1,5-Diaminopentane (cadaverine) is the decarboxylation product of lysine, ornithine yields 1,4-diaminobutane (putrescine).


Synonym(s): N-Boc-cadaverine

Formula: C₁₀H₂₂N₂O₂
MW: 202.30 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00210020
CAS Number: 51644-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR