Phenethyl caffeate

Supplier: BACHEM AMERICAS INC

4027526.0001 4027526.0005
Q-2305.0001BAEA 645.43 CAD
Q-2305.0001BA Q-2305.0005BA
Phenethyl caffeate
Phenethyl caffeate

Synonym(s): CAPE


CAPE is known to have antimitogenic, anticarcinogenic, antiinflammatory, and immunomodulating properties. The molecular basis for these properties seems to be its ability to specifically and potently inhibit the activation of the nuclear transcription factor NF-kappaB.

Formula: C₁₇H₁₆O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00866470
CAS Number: 104594-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR