Antazoline·0.5 H₂SO₄

Supplier: BACHEM AMERICAS INC

4041457.0005 4041457.0025
Q-2115.0005BAEA 232.35 CAD
Q-2115.0005BA Q-2115.0025BA
Antazoline·0.5 H₂SO₄
Antazoline·0.5 H₂SO₄

Sequence: 2-(N-Phenyl-N-benzyl-aminomethyl)-imidazoline · 0.5 H₂SO₄


Antazoline, an imidazoline derivative with antihistaminic properties, has been shown to exert neuroprotection against hypoxia and NMDA toxicity in cerebellar and striatal neuronal cultures through a voltage-dependent blockade of glutamatergic NMDA receptors. Recently, an additional neuroprotective mechanism which functions by decreasing the release of glutamate through inhibition of presynaptic Ca²⁺-channels has been proposed.


Synonym(s): Benzyl-(4,5-dihydro-1H-imidazol-2-ylmethyl)-phenyl-amine · 0.5 H₂SO₄

Formula: 2 C₁₇H₁₉N₃·H₂SO₄
MW: 628.80 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00058146
CAS Number: 91-75-8
EINECS: 202-094-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR