IPTG (isopropyl-β-D-thiogalactopyranoside)
Supplier: BACHEM AMERICAS INC
Synonyms:
IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside
4026913.0005
4026913.0025
Q-2060.0005BAEA
345.56
CAD
Q-2060.0005BA
Q-2060.0025BA
IPTG (isopropyl-β-D-thiogalactopyranoside)
IPTG (Isopropyl-β-D-thiogalactopyranoside)
Sequence: Isopropyl-1-thio-β-D-galactopyranoside (dioxane free)
IPTG is an analog of galactose which is not cleaved by E.coli β-galactosidase. Inducer of the lac operon. β-Galactosidase-positive colonies can be detected by X-Gal (5-Bromo-4-chloroindol-3-yl-β-D-galactoside), M-1190.
Formula:
C₉H₁₈O₅S Storage Temperature: Freezer |
MDL Number:
MFCD00063273 CAS Number: 367-93-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...