4-Aminobenzamidine dihydrochloride
Supplier: BACHEM AMERICAS INC
Synonyms:
4-Aminophenylamidine dihydrochloride, 4-Aminobenzene-1-carboximidamide dihydrochloride
4031123.0005
4031123.0025
Q-1960.0005BAEA
325.67
CAD
Q-1960.0005BA
Q-1960.0025BA
4-Aminobenzamidine dihydrochloride
4-Aminobenzamidine dihydrochloride
Sequence: p-Aminobenzamidine · 2 HCl
Competitive inhibitor of trypsin, plasmin and thrombin. CAS Number (net): 3858-83-1.
Formula:
C₇H₁₁Cl₂N₃ MW: 208.09 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00013001 CAS Number: 2498-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...