EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)
Supplier: BACHEM AMERICAS INC
Synonyms:
N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine monohydrochloride, N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide monohydrochloride, 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDC hydrochloride, EDC, EDAC HCl, EDAC hydrochloride, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, WSC HCl, EDC HCl
Synonym(s): EDAC · HCl#EDC · HCl#N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide · HCl#Water-soluble Carbodiimide#WSC
A water-soluble coupling reagent for solution synthesis. WSCD has also been employed in SPPS in aqueous systems.
Formula:
C₈H₁₇N₃·HCl MW: 191.7 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00012503 CAS Number: 25952-53-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...