2,5-dioxopyrrolidin-1-yldodecanoate

Supplier: BACHEM AMERICAS INC

Synonyms: Lauric Acid-N-Hydroxy-Succinimide Ester, Lauric acid-OSu, 2,5-Dioxopyrrolidin-1-yl dodecanoate

4026254.0005 4026254.0025
Q-1920.0005BAEA 619.6 CAD
Q-1920.0005BA Q-1920.0025BA
2,5-dioxopyrrolidin-1-yldodecanoate
2,5-dioxopyrrolidin-1-yldodecanoate

Sequence: Lauric acid-OSu
Synonym(s): Dodecanoic acid-OSu


For DOTA(OtBu)₃ see Q-2525.


Suitable for the synthesis of lauroyl derivatives of amines and thiols. Reagent for the modification of peptides to increase their lipophilicity.

Formula: C₁₆H₂₇NO₄
MW: 297.395 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00050401
CAS Number: 14565-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR