Agmatine sulfate
Supplier: BACHEM AMERICAS INC
4026181.0005
4026181.0001
Q-1880.0005BAEA
828.77
CAD
Q-1880.0005BA
Q-1880.0001BA
Agmatine sulfate
Agmatine sulfate
1-Amino-4-guanidinobutane · sulfate
CAS: 2482-00-0
Formula: C5H14N4 · H2SO4
M.W: 228.27
Storage temp.: +5 ± 3 °C
Formula:
C₅H₁₄N₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD00013109 CAS Number: 2482-00-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...