D-(-)-Lactic acid
Supplier: BACHEM AMERICAS INC
4003254.0250
4003254.1000
Q-1870.0250BAEA
309.81
CAD
Q-1870.0250BA
Q-1870.1000BA
D-(-)-Lactic acid
D(-)-Lactic acid
Sequence: D-Lactic acid
Useful chiral synthons, building blocks for depsipeptides.
Synonym(s): (R)-2-Hydroxy-propionic acid#H-D-Lac-OH
Formula:
C₃H₆O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD00068313 CAS Number: 10326-41-7 EINECS: 233-713-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...