7-Methoxycoumarin-4-acetic acid
Supplier: BACHEM AMERICAS INC
Synonyms:
7-Methoxy-2-oxo-2H-1-benzopyran-4-acetic acid, Mca, 2-(7-Methoxy-2-oxo-2H-chromen-4-yl)acetic acid, (7-Methoxycoumarin-4-yl)acetic acid
Sequence: (7-Methoxycoumarin-4-yl)acetic acid
Synonym(s): Mca-OH
Bachem also offers the coumarin derivative Fmoc-3-(7-methoxy-coumarin-4-yl)-Ala-OH (B-3740), the (7-Hydroxycoumarin-4-yl)ethylglycine derivatives B-4205 and F-4270, and a range of Nε-labeled lysine derivatives (please see the subfamily "Nε-Labeled Lysine").
Formula:
C₁₂H₁₀O₅ MW: 234.207 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00009774 CAS Number: 62935-72-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...