9-Fluorenylmethyl carbamate
Supplier: BACHEM AMERICAS INC
Synonyms:
Carbamic Acid 9-Fluorenylmethyl Ester, Fmoc-NH2, (9H-Fluoren-9-ylmethoxy)carboxamide
4012755.0005
4012755.0025
Q-1830.0005BAEA
413.58
CAD
Q-1830.0005BA
Q-1830.0025BA
9-Fluorenylmethyl carbamate
9-Fluorenylmethyl carbamate
Sequence: Fmoc-NH₂
Synonym(s): Fmoc-amide
For DOTA(OtBu)₃ see Q-2525.
9-Fluorenylmethyl carbamate is an important educt for the synthesis of Fmoc-protected amide linkers.
Formula:
C₁₅H₁₃NO₂ MW: 239.27 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00237376 CAS Number: 84418-43-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...