N-Hydroxysuccinimide

Supplier: BACHEM AMERICAS INC

4025878.0100 4025878.0500
Q-1800.0100BAEA 238.17 CAD
Q-1800.0100BA Q-1800.0500BA
N-Hydroxysuccinimide
N-Hydroxysuccinimide

Sequence: N-Hydroxysuccinimide
Synonym(s): HOSu


Educt for the synthesis of N-hydroxysuccinimide esters of N-substituted amino acids and peptides. Additive for peptide synthesis by the azide fragment condensation on a soluble polymer support. Chiral stabilizer for peptide coupling reactions.

Formula: C₄H₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD00005516
CAS Number: 6066-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR