D-Biotin-N-succinimidyl ester

Supplier: BACHEM AMERICAS INC

4014225.0250 4014225.1000
Q-1760.0250BAEA 413.45 CAD
Q-1760.0250BA Q-1760.1000BA
D-Biotin-N-succinimidyl ester
D-Biotin-N-succinimidyl ester

Sequence: D-Biotinyl-OSu
Synonym(s): BNHS#Biotin-NHS


For DOTA(OtBu)₃ see Q-2525.


The activated biotin derivative Biotin-OSu is used for labeling the amino groups of amino acids, peptides, and proteins. When conducting the biotinylation at pH 6.5, N-terminal amino groups can be selectively tagged in the presence of lysine.

Formula: C₁₄H₁₉N₃O₅S
Storage Temperature: Refrigerator
MDL Number: MFCD00078531
CAS Number: 35013-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR