3-(Tritylthio)propanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: 3-Tritylsulfanyl-propionic acid, S-Trityl-3-mercaptopropionic acid, 3-Tritylsulphanyl-propionic acid, 3-(Tritylthio)propionic acid, Mpa(Trt)

4012270.0001 4012270.0005
Q-1740.0001BAEA 206.51 CAD
Q-1740.0001BA Q-1740.0005BA
3-(Tritylthio)propanoic acid
3-(Tritylthio)propanoic acid

Sequence: 3-Tritylsulfanyl-propionic acid
Synonym(s): Mpr(Trt)-OH#Mpa(Trt)-OH#S-Trityl-deamino-cysteine


β-Mercaptopropionic acid (deamino-cysteine, Mpr, Mpa) is often incorporated in analogs of bioactive peptides as a replacement of an N-terminal Cys.


S-Protected derivative for introducing Mpr during Fmoc-SPPS.

Formula: C₂₂H₂₀O₂S
MW: 348.46 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00237291
CAS Number: 27144-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR