2-​Acetamido-​2-​deoxy-​​β-​D-​glucopyranosyl L-​asparagine

Supplier: BACHEM AMERICAS INC

Synonyms: β-D-GlcNAc-(1→N)-Asn, Ng-(2-Acetamido-2-deoxy-b-D-gluco-pyranosyl)-L-asparagine, 2-Acetamido-1-β-(L-aspartamido)-1 ,2-dideoxy-D-glucose, H-Asn(GlcNAc-β-D)-OH

4027708.0250 4027708.1000
Q-1725.0250BAEA 2118.32 CAD
Q-1725.0250BA Q-1725.1000BA
2-​Acetamido-​2-​deoxy-​​β-​D-​glucopyranosyl L-​asparagine
2-​Acetamido-​2-​deoxy-​​β-​D-​glucopyranosyl L-​asparagine

Sequence: H-Asn(GlcNAc-β-D)-OH


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".


Synonym(s): AADG#Aspartylglucosamine#N-γ-(2-Acetamido-2-deoxy-β-D-gluco-pyranosyl)-L-asparagine

Formula: C₁₂H₂₁N₃O₈
MW: 335.31 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02259695
CAS Number: 2776-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR