L-1,2,3,4-Tetrahydronorharman-3-carboxylic acid

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2,3,4,9-Tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic acid

4016480.0001 4016480.0005
Q-1700.0001BAEA 180.91 CAD
Q-1700.0001BA Q-1700.0005BA
L-1,2,3,4-Tetrahydronorharman-3-carboxylic acid
L-1,2,3,4-Tetrahydronorharman-3-carboxylic acid

Sequence: L-1,2,3,4-Tetrahydronorharman-3-carboxylic acid


Tpi, a constrained Trp analog, is also called 1,2,3,4-Tetrahydro-β-carboline-3-carboxylic acid.


Synonym(s): H-Tpi-OH#L-1,2,3,4-Tetrahydro-β-carboline-3-carboxylic acid#L-Tryptoline-3-carboxylic acid#L-1,2,3,4-Tetrahydro-9H-pyrido[3,4-b]indole-3-carboxylic acid

Formula: C₁₂H₁₂N₂O₂
MW: 216.24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00272641
CAS Number: 42438-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR