Potassium-1-carboxyvinyl hydrogenphosphate

Supplier: BACHEM AMERICAS INC

Synonyms: Phosphoenolpyruvate potassium salt, Potassium 1-carboxyvinyl hydrogenphosphate, Phosphoenolpyruvic acid monopotassium salt, Phosphoenolpyruvic acid potassium

4014027.0005 4014027.0025
Q-1640.0005BAEA 549.38 CAD
Q-1640.0005BA Q-1640.0025BA
Potassium-1-carboxyvinyl hydrogenphosphate
Potassium-1-carboxyvinyl hydrogenphosphate

Sequence: Phosphoenolpyruvate potassium salt
Synonym(s): PEP-K#Potassium phosphoenolpyruvate#2-Phosphonooxy-acrylic acid potassium salt


Substrate for many kinases such as pyruvate kinase and phosphoenolpyruvate carboxykinase.

Formula: C₃H₄KO₆P
Storage Temperature: Refrigerator
MDL Number: MFCD00044476
CAS Number: 4265-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR