Mal-sac-HNSA

Supplier: BACHEM AMERICAS INC

Synonyms: ε-Maleimidocaproic acid-(2-nitro-4-sulfo)-phenyl ester sodium salt, ε-Maleimidocaproic acid-(2-nitro-4-sulpho)-phenyl ester sodium salt

4013577.0100 4013577.0500
Q-1615.0100BAEA 387.21 CAD
Q-1615.0100BA Q-1615.0500BA
Mal-sac-HNSA
Mal-sac-HNSA

Sequence: ε-Maleimidocaproic acid-(2-nitro-4-sulfo)-phenyl ester · sodium salt
Synonym(s): mal-sac-HNSA


mal-sac-HNSA, an often used freely water-soluble, heterobifunctional crosslinking reagent for conjugation of sulfhydryl (cysteine)-containing peptides to carrier proteins. The reaction of mal-sac-HNSA with amino groups can be monitored spectrophotometrically due to the concomitant release of the dianion of 1-hydroxy-2-nitro-4-benzenesulfonic acid.

Formula: C₁₆H₁₅N₂NaO₉S
MW: 435.359 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00236937
CAS Number: 101554-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR