(R)-(-)-2-Oxothiazolidine-4-carboxylic acid

Supplier: BACHEM AMERICAS INC

Synonyms: (R)-2-Oxothiazolidine-4-carboxylic acid, Procysteine

4012415.0001 4012415.0005
Q-1535.0001BAEA 180.91 CAD
Q-1535.0001BA Q-1535.0005BA
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid

Sequence: L-2-Oxothiazolidine-4-carboxylic acid


Thiazolidine-4-carboxylic acid (Thz) can be inserted as a Pro equivalent (thiaproline) as well as a masked cysteine. Contrary to Cys derivatives, Thz derivatives can be activated and coupled with minimal racemization.


Synonym(s): OTC#Procysteine#OTZ#L-Thiazolidin-2-one-4-carboxylic acid

Formula: C₄H₅NO₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00066092
CAS Number: 19771-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR