Toluene-α-sulfonyl fluoride
Supplier: BACHEM AMERICAS INC
Synonyms:
PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulfonyl fluoride, α-Toluenesulphonyl fluoride, Phenylmethanesulfonyl fluoride, Phenylmethylsulfonyl fluoride
4000472.0025
4000472.0100
Q-1395.0025BAEA
325.67
CAD
Q-1395.0025BA
Q-1395.0100BA
Toluene-α-sulfonyl fluoride
Toluene-α-sulphonyl fluoride
Sequence: Phenylmethylsulfonyl fluoride
Synonym(s): Pms-F#PMSF
PMSF inhibits serine proteases such as trypsin and chymotrypsin, cysteine proteases such as papain, and acetylcholinesterase.
Formula:
C₇H₇FO₂S MW: 174.2 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00007424 CAS Number: 329-98-6 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...