DL-3-Phenyllactic acid

Supplier: BACHEM AMERICAS INC

4000118.0025 4000118.0100
Q-1380.0025BAEA 317.52 CAD
Q-1380.0025BA Q-1380.0100BA
DL-3-Phenyllactic acid
DL-3-Phenyllactic acid

Sequence: DL-β-Phenyllactic acid


Useful chiral synthons, building blocks for depsipeptides.


Synonym(s): (RS)-2-Hydroxy-3-phenyl-propionic acid

Formula: C₉H₁₀O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00065928
CAS Number: 828-01-3
EINECS: 212-580-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR